missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Heavy Chain Antibody (IGHA/7567R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20706JF635
This item is not returnable.
View return policy
Description
IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IgA Heavy Chain | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| IGHA/7567R | |
| 50mM Sodium Borate | |
| Full-length native protein (purified) corresponding to Human IgA | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction