missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (IGHD/8228R), Novus Biologicals™
Rabbit Monoclonal Antibody
298.00 € - 519.00 €
Specifications
| Antigen | IgD |
|---|---|
| Clone | IGHD/8228R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694724
|
Novus Biologicals
NBP3-20663-20ug |
20 μg |
298.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600454
|
Novus Biologicals
NBP3-20663-100ug |
100 μg |
519.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| IgD | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| B Cell Development and Differentiation Markers, Immunology | |
| 1mM PBS with 0.05% BSA | |
| 3495 | |
| IgG κ | |
| Protein A or G purified |
| IGHD/8228R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Recombinant fragment (around aa1-200) of human IgD protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title