missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgD Monoclonal Antibody (IA6-2), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ H026T03Y04-A
This item is not returnable.
View return policy
Description
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Specifications
| Human IgD | |
| Monoclonal | |
| 0.2 μg/Test | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01880 | |
| IGHD | |
| Affinity chromatography | |
| RUO | |
| 3495 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG2a κ |
| Flow Cytometry | |
| IA6-2 | |
| NovaFluor Yellow 660 | |
| IGHD | |
| IGHD; immunoglobulin heavy constant delta | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction