missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgD Monoclonal Antibody (IA6-2), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | Human IgD |
|---|---|
| Clone | IA6-2 |
| Concentration | 0.2 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055375
|
Invitrogen™
H026T03R06-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055591
|
Invitrogen™
H026T02R06-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow CytometrySpecifications
| Human IgD | |
| 0.2 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01880 | |
| 3495 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| IA6-2 | |
| Flow Cytometry | |
| NovaFluor Red 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IGHD; immunoglobulin heavy constant delta | |
| IgG2a κ | |
| Affinity chromatography | |
| IGHD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title