missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgE Monoclonal Antibody (Ige21), NovaFluor™ Red 725, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

185.00 € - 392.00 €

Specifications

Antigen Human IgE
Clone Ige21
Concentration 0.2 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30056186
View Documents
Invitrogen™
H077T03R05-A
100 Tests
392.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30054832
View Documents
Invitrogen™
H077T02R05-A
25 Tests
185.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

Human IgE
0.2 μg/Test
Monoclonal
Mouse
Human
P01854
3497
Primary
4°C, store in dark, DO NOT FREEZE!
Ige21
Flow Cytometry
NovaFluor Red 725
RUO
PBS with 0.09% sodium azide; pH 7.2
HCAK1; IgA1; IgG3; IGHA1; IGHA2; IGHG1; IGHG2; IGHG3; IGHG4; IGKC; IGKCD; IGLC; IGLC1; Immunoglobulin E; immunoglobulin heavy constant alpha 1; immunoglobulin heavy constant alpha 2 (A2m marker); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); immunoglobulin kappa constant; immunoglobulin lambda constant 1; ImmunoglobulinE; Km
IgG1 κ
Affinity chromatography
IGHE
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.