missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgE Monoclonal Antibody (Ige21), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ H077T02Y07-A
This item is not returnable.
View return policy
Description
IgE is one of the five classes of immunoglobulins found in mammals, characterized by its monomeric structure consisting of two heavy (epsilon) chains with four Ig-like constant domains and two light chains. Although IgE comprises only a small portion of the total immunoglobulin in serum, it plays a crucial role in type I hypersensitivity reactions, such as hay fever, asthma, hives, and anaphylactic shock. IgE is produced by terminally differentiated plasma B cells and basophils. It binds to Fc epsilon receptors on mast cells and basophils, leading to the release of histamine and other molecules that promote allergic responses. Crosslinking of IgE bound to these receptors induces degranulation of mast cells and basophils, resulting in allergic manifestations. In addition to its role in allergic responses, IgE is functionally important for immunity against parasites, highlighting its dual role in both protective and pathological immune processes.
Specifications
| Human IgE | |
| Monoclonal | |
| 0.2 μg/Test | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01854 | |
| IGHE | |
| Affinity chromatography | |
| RUO | |
| 3497 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG1 κ |
| Flow Cytometry | |
| Ige21 | |
| NovaFluor Yellow 730 | |
| IGHE | |
| HCAK1; IgA1; IgG3; IGHA1; IGHA2; IGHG1; IGHG2; IGHG3; IGHG4; IGKC; IGKCD; IGLC; IGLC1; Immunoglobulin E; immunoglobulin heavy constant alpha 1; immunoglobulin heavy constant alpha 2 (A2m marker); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); immunoglobulin kappa constant; immunoglobulin lambda constant 1; ImmunoglobulinE; Km | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction