Learn More
IGF2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547946
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant human IGF-II is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IGF2 Polyclonal antibody specifically detects IGF2 in Mouse samples. It is validated for Immunohistochemistry (Frozen), Neutralization, Western Blot.Specifications
IGF2 | |
Polyclonal | |
Unconjugated | |
IGF2 | |
AL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16002 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P09535 | |
IGF2 | |
E. coli-derived recombinant mouse IGF-II Ala25-Glu91 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |