Learn More
IGFBP-1 Monoclonal Antibody (33627.11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA124775
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-24775 detects IGFBP1 from human samples. MA1-24775 has been successfully used in neutralization, ELISA , and Western blot procedures. The Neutralization Dose50 (ND50) for this antibody is 10-40 μg/mL in the presence of approximately 5 μg/mL of recombit human IGFBP1 and approximately 6 ng/mL of recombit human IGF1, using human MCF7 cells. The MA1-24775 immunogen is recombit, full length human protein.
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.Specifications
IGFBP1 | |
Monoclonal | |
0.5 mg/mL | |
IGFBP1 | |
AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1 | |
Mouse | |
Protein G | |
RUO | |
3484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Neutralization, Western Blot | |
33627.11 | |
Unconjugated | |
P08833 | |
IGFBP1 | |
Recombit, full length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |