Learn More
IGFBP-1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547102
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant human IGFBP-1 and less than 1% cross-reactivity with recombinant mouse (rm) IGFBP-2, rmIGFBP-3, rmIGFBP-5, rmIGFBP-6, and rmIGFBP-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
IGFBP-1 Polyclonal antibody specifically detects IGFBP-1 in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
IGFBP-1 | |
Polyclonal | |
Unconjugated | |
IGFBP1 | |
AFBP; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; Binding protein 25; Binding protein 26; Binding protein 28; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP 1; hIGFBP1; hIGFBP-1; IBP 1; IBP1; IBP-1; IGF BP25; IGFBA; IGF-binding protein 1; IGFBP; IGFBP 1; Igfbp1; Igfbp-1; IGF-BP25; Insulin like growth factor binding protein; insulin like growth factor binding protein 1; insulin-like growth factor binding protein 1; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); insulin-like growth factor-binding protein 1; Placental protein 12; PP 12; PP12 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16006 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P47876 | |
IGFBP1 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-1 Ala26-Asn272 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |