Learn More
IGFBP-2 Antibody (CL13565) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24569AF350
Product Code. 30301113
  • 737.00 € / 0.10mL
Description
IGFBP-2 Monoclonal antibody specifically detects IGFBP-2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IGFBP-2
Monoclonal
Alexa Fluor 350
BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2
This antibody was developed using a synthetic peptide derived from P18065, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cytokines and Growth Factors, Signal Transduction
3485
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL13565
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified