missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGFBP6 Monoclonal Antibody (122312)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561864
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
IGF-BPs control the distribution, function and activity of IGFs in various cell tissues and body fluids. IGF-BP6, which specifically inhibits IGF-II actions, is produced by bone cells and is the major IGF-BP present in cerebrospinal fluid. IGF-BP6 has been shown to inhibit IGF-II-dependent cancers such as neuroblastoma, colon cancer and rhabdomyosarcoma.
Specifications
| IGFBP6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P47880 | |
| IGFBP6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-6, Gly24-Gly238. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| 122312 | |
| Unconjugated | |
| IGFBP6 | |
| IBP6; IBP-6; IGF binding protein 6; IGF-binding protein 6; IGFBP; IGFBP6; IGF-BP6; Igfbp-6; Insulin like growth factor binding protein; insulin like growth factor binding protein 6; insulin-like growth factor binding protein 6; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 6 PRECURSOR (IGFBP-6) (IBP-6) (IGF-BINDING PROTEIN 6); insulin-like growth factor-binding protein 6; RBP6A | |
| Rat | |
| Protein A/G | |
| RUO | |
| 16012 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction