missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Antibody (E20-V), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08196-100ul
This item is not returnable.
View return policy
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| IgG | |
| Monoclonal | |
| Unconjugated | |
| 20 mM Tris-HCl, pH 8.0, 20 mg/ml BSA | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| E20-V | |
| Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300 | |
| Immunoglobulin G, ImmunoglobulinG | |
| Peptide derived from internal domain of human IgG-1 chain C region. Antibody recognizes the epitope betweenVal167 - Val185. | |
| 100 μg | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| 3500 | |
| Store at 4°C. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion