missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Light Chain Antibody (IVA285-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62190
This item is not returnable.
View return policy
Description
IgG Light Chain Monoclonal antibody specifically detects IgG Light Chain in Bovine, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| IgG Light Chain | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bovine, Sheep | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| IVA285-1 | |
| Western Blot 0.5-1 μg/ml, Flow Cytometry 1-4 μg/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| HCAK1;IGK@;Igk-C;IGKCD;kappa;Km | |
| Fraction of Bovine immunoglobulin. | |
| 0.1 mg | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction