missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG4 (Ig Heavy Constant Gamma 4) (G4m Marker) Rabbit, Clone: IGHG4/2042R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 3503-RBM2-P0
This item is not returnable.
View return policy
Specifications
| IgG4 (Ig Heavy Constant Gamma 4) (G4m Marker) | |
| Cytoplasmic | |
| IGHG4/2042R | |
| Unconjugated | |
| Rabbit | |
| 150kDa | |
| Protein A column purification. | |
| RUO | |
| 3503 | |
| IgG |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human IGHG4 protein | |
| Tonsil | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction