Learn More
IgM Antibody (II/41), PE, Novus Biologicals™
Rat Monoclonal Secondary Antibody
Brand:  Novus Biologicals NBP1-43763-0.1MG
 View more versions of this product
Product Code. 18286233
  • 418.00 € / 0.10mg
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgM
Monoclonal
0.2 mg/mL
Flow Cytometry
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Rat
Affinity Purified
RUO
Secondary
Mouse
IgG2a κ
Flow Cytometry
II/41
PE
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
IGHM
The immunogen for this antibody was IgM.
0.1 mg
B Cell Development and Differentiation Markers, Immunology
3507
Store at 4C. Do not freeze.