Learn More
IGSF11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561231
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PPKCSSAKAF HTEISSSDNN TLTSSNAYNS RYWSNNPKVH RNTESVSHFS DLGQSFSFHS GNANIPSIYA NGTHLVPGQH KTLVVTANRG SSPQVMSRSN GSVSRKP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
Functions as a cell adhesion molecule through homophilic interaction. Stimulates cell growth.Specifications
IGSF11 | |
Polyclonal | |
Unconjugated | |
Igsf11 | |
1700025L02Rik; brain and testis-specific immunoglobin superfamily protein; brain and testis-specific immunoglobulin superfamily protein; BTIGSF; Bt-IGSF; cancer/testis antigen 119; CT119; CXADR like 1; CXADRL1; Igsf11; Igsf13; Immunoglobulin superfamily member 11; immunoglobulin superfamily, member 11; immunoglobulin superfamily, member 13; V-set and immunoglobulin domain containing 3; V-set and immunoglobulin domain-containing protein 3; VSIG3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152404 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5DX21 | |
Igsf11 | |
Recombinant protein corresponding to Human IGSF11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only