missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ikaros/IKZF1 Antibody (S08-5E4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19660-50ul
This item is not returnable.
View return policy
Description
Ikaros/IKZF1 Monoclonal antibody specifically detects Ikaros/IKZF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Ikaros/IKZF1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin | |
| DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6 | |
| A synthetic peptide of human Ikaros/IKZF1 (Uniprot # Q13422) | |
| 50 μg | |
| Innate Immunity | |
| 10320 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S08-5E4 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction