Learn More
Invitrogen™ IkB zeta Recombinant Rat Monoclonal Antibody (LK2NAP)
Rat Recombinant Monoclonal Antibody
Brand: Invitrogen™ 740065T20UG
Description
Recombinant rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
I-kappa-B-zeta (also known as MAIL and INAP) is an ankyrin-repeat-containing nuclear protein that is highly homologous to the IkappaB family member Bcl-3. Transcription of IkappaBzeta is upregulated by stimulation with TLR ligands, IL-1 and IL-6 in cultured B-lymphocytes and monocytes/macrophages, but only faintly so in T-lymphocytes, fibroblasts, and endothelial cells. IkappaB-zeta preferentially associates with the NF-kappaB subunit p50 rather than p65 and recombinant IkappaB-zeta proteins inhibit the DNA binding of the p65/p50 heterodimer and the p50/p50 homodimer.
Specifications
| IkB zeta | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9BYH8, Q9EST8 | |
| NFKBIZ | |
| Affinity chromatography | |
| RUO | |
| 304005, 64332, 80859 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| LK2NAP | |
| Unconjugated | |
| NFKBIZ | |
| AA408868; FLJ30225; FLJ34463; Ikappa B-zeta variant 3; ikappaBzeta; I-kappa-B-zeta; IkappaB-zeta; IKBZ; ikB-zeta; IL-1 inducible nuclear ankyrin-repeat protein; INAP; Mail; molecule possessing ankyrin repeats induced by lipopolysaccharide; molecule possessing ankyrin-repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta; NFKB inhibitor zeta; NFKBIZ; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta; OTTHUMP00000214340; RGD1310834 | |
| Rat | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.