missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-1 alpha/IL-1F1 Antibody Pair [HRP]

Product Code. p-7183181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18205978 15 Plates Pack of 15
18625620 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18205978 Supplier Novus Biologicals™ Supplier No. NBP27952215PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Mouse IL-1 alpha/IL-1F1.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 6.25 pg/mL
Assay Sensitivity 6.25-400 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse IL-1 alpha/IL-1F1.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha
Gene ID (Entrez) 3552
Gene Symbol IL1A
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 30 ng of recombinant Mouse IL-1 alpha/IL-1F1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Mouse IL-1 alpha/IL-1F1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-IL-1 alpha/IL-1F1 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
    Storage Requirements Storage is content dependent.
    Target IL-1 alpha/IL-1F1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 400 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.