missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-1 RAcP/IL-1 R3 Antibody (nidanilimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27990-100ug
This item is not returnable.
View return policy
Description
IL-1 RAcP/IL-1 R3 Monoclonal antibody specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-1 RAcP/IL-1 R3 | |
| Monoclonal | |
| Unconjugated | |
| Q9NPH3 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| nidanilimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta | |
| IL-1RAP/ IL-1R3 | |
| 100 μg | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| 3556 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction