missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11366-0.1mg
This item is not returnable.
View return policy
Description
IL-13 Polyclonal antibody specifically detects IL-13 in Canine samples. It is validated for Western Blot, ELISA
Specifications
| IL-13 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Canine | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| Recombinant canine IL-13 | |
| 0.1 mg | |
| Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3596 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction