missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-13R alpha 1 Antibody Pair [HRP]

Product Code. 18659450 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18659450

Brand: Novus Biologicals™ NBP2793265Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-13R alpha 1.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-13R alpha 1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4
Gene ID (Entrez) 3597
Gene Symbol IL13RA1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 80 ng of recombinant IL-13R alpha 1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-IL-13R alpha 1 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL of mouse anti-IL-13R alpha 1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 5 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target IL-13R alpha 1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1.0 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A six-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.