missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13R alpha 2 Antibody (MM0367-11S23) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
206.00 € - 507.00 €
Specifications
| Antigen | IL-13R alpha 2 |
|---|---|
| Clone | MM0367-11S23 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18077576
|
Novus Biologicals
NBP2-11663-0.025MG |
0.025 mg |
206.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18051947
|
Novus Biologicals
NBP2-11663 |
0.1 mg | |||||||
Description
IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Human samples. It is validated for Western Blot.Specifications
| IL-13R alpha 2 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| Q14627 | |
| 3598 | |
| IgG1 | |
| Protein G purified |
| MM0367-11S23 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
| IL13RA2 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title