missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Rabbit anti-Human, Clone: 111, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89449-50UL
This item is not returnable.
View return policy
Description
IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| IL-15 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 111 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| IL-15MGC9721, interleukin 15, interleukin-15 | |
| Recombinant Human IL-15 Protein | |
| 50 μg | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3600 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction