missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21240-25ul
This item is not returnable.
View return policy
Description
IL-16 Polyclonal antibody specifically detects IL-16 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IL-16 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QRARSFPLTRSQSCETKLLDEKTSKLYSISSQVSSAVMKSLLCLPSSISCAQTPCIPKEGASPTSSSNEDSAANGSAETSALDTGFSLNLSELREYTEGLTEAKEDDDG | |
| 25 μg | |
| Biologically Active Proteins, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity | |
| 3603 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction