missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (4H450) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
210.00 € - 585.00 €
Specifications
| Antigen | IL-17F |
|---|---|
| Clone | 4H450 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.2 - 2 ug/ml |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18127182
|
Novus Biologicals
NBP2-27319SS |
0.025 mg |
210.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717233
|
Novus Biologicals
NBP2-27319 |
585.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Western Blot.Specifications
| IL-17F | |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cytokine Research | |
| Q96PD4 | |
| 112744 | |
| Full-length recombinant MRkIPkVGHT FFQkPESCPP VPGGSMkLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGkEDISMN SVPIQQETLV VRRkHQGCSV SFQLEkVLVT VGCTCVTPVI HHVQ | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 4H450 | |
| Western Blot 0.2 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24 | |
| IL17F | |
| IgG1 κ | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title