missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1ra/IL-1F3/IL1RN Antibody (SR1317), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21689-50ul
This item is not returnable.
View return policy
Description
IL-1ra/IL-1F3/IL1RN Monoclonal antibody specifically detects IL-1ra/IL-1F3/IL1RN in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Serum
Specifications
| IL-1ra/IL-1F3/IL1RN | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| SR1317 | |
| Western Blot 1:5000-1:20000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50, Immunocytochemistry, Serum 1:50-1:200 | |
| DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| A synthesized peptide derived from human IL-1ra/IL-1F3/IL1RN (Uniprot #: P18510) | |
| 50 μg | |
| Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3557 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction