missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-1RAcP Recombinant Human Monoclonal Antibody (2G7)
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560732
This item is not returnable.
View return policy
Description
IL-1RAcP Recombinant Monoclonal Antibody for ELISA
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
Specifications
| IL-1RAcP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9NPH3 | |
| IL1RAP | |
| Recombinant Human IL1RAP protein. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| ELISA | |
| 2G7 | |
| Unconjugated | |
| IL1RAP | |
| 6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1R-3; IL1RAcP; IL-1RAcp; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 3556 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction