Learn More
IL-2 R beta Antibody (TM-b1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP1-43355-0.5mg
 View more versions of this product
Product Code. 18740594
  • 617.00 € / 0.50mg
Description
IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-2 R beta
Monoclonal
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess
CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Rat
Affinity Purified
RUO
3560
Store at 4C. Do not freeze.
Flow Cytometry, Immunoprecipitation, Blocking Assay
TM-b1
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
IL2RB
The immunogen for this antibody was Purified CD122.
0.5 mg
Primary
Mouse
IgG2b