missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 R beta Antibody (TM-b1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43355-0.025MG
This item is not returnable.
View return policy
Description
IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Block/Neutralize.
Specifications
| IL-2 R beta | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| 3560 | |
| Store at 4C. Do not freeze. |
| Flow Cytometry, Immunoprecipitation, Blocking Assay | |
| TM-b1 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| IL2RB | |
| The immunogen for this antibody was Purified CD122. | |
| 0.025 mg | |
| Primary | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction