missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-2 Recombinant Rabbit Monoclonal Antibody (449), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
109.00 € - 218.00 €
Specifications
| Antigen | IL-2 |
|---|---|
| Clone | 449 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019933
|
Invitrogen™
MA552435 |
25 Tests |
109.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019642
![]() |
Invitrogen™
MA552434 |
100 Tests |
218.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
IL-2 Recombinant Monoclonal antibody specifically detects IL-2 in Mouse samples. It is validated for Flow CytometryTekniske data
| IL-2 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
| Recombinant Mouse Interleukin-2/IL-2 Protein (Ala21-Gln169). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IL2 |
| 449 | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Mouse | |
| P04351 | |
| 16183 | |
| IgG | |
| Protein A | |
| Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel