Learn More
Novus Biologicals™ IL-20R alpha Antibody (Cheng Kung U. patent anti-IL-20R1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28332-50ug
 View more versions of this product
Product Code. 30045766
  • 213.00 € / 50µg
Description
IL-20R alpha Monoclonal antibody specifically detects IL-20R alpha in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-20R alpha
Monoclonal
Unconjugated
Q9UHF4
Human
Protein A purified
RUO
53832
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Cheng Kung U. patent anti-IL-20R1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7
IL-20Ra
50 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.