Learn More
Invitrogen™ IL-23 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559562
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term LY2525623.
IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 that is disulfide-linked with to two subunits, a p19 subunit unique to IL-23, and a p40 subunit that is shared with IL-12. IL-23 interacts with IL-12Rbeta1 and a beta2-like receptor subunit with STAT4 binding domain, IL-23R. IL-23 is secreted by activated mouse and human dendritic cells and is involved in the inflammation process and associated with autoimmune diseases. Mouse IL-23 was found not to induce significant amounts of IFN-gamma, induces strong proliferation of memory T cells (but not naive T cells), and activate mouse memory T cells to produce the proinflammatory cytokine IL-17. On the other hand, human IL-23 induces proliferation of memory T cells, induces moderate levels of IFN-gamma, and is produced by naive and memory T cells. IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance, and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines. IL-23 interacts with the receptors IL-12R beta1 and IL-23R to activate the Jak-Stat signaling cascade. IL-23 has been shown to possess potent anti-tumor and anti-metastatic activity in mouse models of cancer, suggesting a potential role for IL-23 in therapeutic treatment of cancer.
Specifications
| IL-23 Biosimilar | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.