missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-23R Antibody (15N6C6), Novus Biologicals™
Mouse Monoclonal Antibody
226.00 € - 381.00 €
Specifications
| Antigen | IL-23R |
|---|---|
| Clone | 15N6C6 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1 - 3 ug/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18143722
|
Novus Biologicals
NBP2-27091SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758813
|
Novus Biologicals
NBP2-27091 |
381.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
IL-23R Monoclonal specifically detects IL-23R in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-23R | |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| Q5VWK5 | |
| 149233 | |
| A synthetic peptide corresponding to amino acids 288-315 of hIL-23 receptor was used as immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 15N6C6 | |
| Western Blot 1 - 3 ug/mL, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| IL23R | |
| IgG1 κ | |
| Protein G purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in Human, Chimpanzee and Orangutan, 89% in Mouse, Pig, Dog and Rhesus Monkey, and 84% homologous in Rat, Horse and Bovine. There are multiple isoforms of IL-23 receptor. Researchers are encouraged to use a tool like BLAST to determine the suitability of this antibody for their testing. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title