missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-32 Antibody (SR1365), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22353-50ul
This item is not returnable.
View return policy
Description
IL-32 Monoclonal antibody specifically detects IL-32 in Human samples. It is validated for Western Blot
Specifications
| IL-32 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1365 | |
| Western Blot 1:500-1:2000 | |
| IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| A synthesized peptide derived from human IL-32 (Uniprot #: P24001) | |
| 50 μg | |
| Cancer, Signal Transduction | |
| 9235 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction