missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-33 Antibody (CL13438) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24558C
This item is not returnable.
View return policy
Description
IL-33 Monoclonal antibody specifically detects IL-33 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| IL-33 | |
| Monoclonal | |
| DyLight 650 | |
| C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
| This antibody was developed using a recombinant protein derived from O95760, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Immunology, Inflammation, Innate Immunity, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 90865 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL13438 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction