missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 351.00 €
Specifications
| Antigen | IL-27/IL-35 EBI3 Subunit |
|---|---|
| Concentration | 0.5 mg/ml |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18048786
|
Novus Biologicals
NBP2-03944SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18739763
|
Novus Biologicals
NBP2-03944 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBI3 Polyclonal specifically detects EBI3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-27/IL-35 EBI3 Subunit | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| EBI3 | |
| IgG | |
| Affinity Purified | |
| Reacts with Human, Mouse and Rat. The amino acid sequence used as immunogen is 100% homologous in mouse (isoforms CRA_a and CRA_b) and rat and 72% homologous in human, dog and rhesus monkey. |
| 0.5 mg/ml | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| NP_056581.1 | |
| 10148 | |
| A portion of amino acids 180-228 of mouse EBI3 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title