missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6R alpha Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
213.00 € - 507.00 €
Specifications
| Antigen | IL-6R alpha |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Flow Cytometry 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229160
|
Novus Biologicals
NBP3-33510-100ul |
100 μL |
507.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227027
|
Novus Biologicals
NBP3-33510-20ul |
20 μL |
213.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Mouse samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| IL-6R alpha | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-357 of mouse IL-6R alpha (NP_034689.2).,, Sequence:, LVLGSCRALEVANGTVTSLPGATVTLICPGKEAAGNVTIHWVYSGSQNREWTTTGNTLVLRDVQLSDTGDYLCSLNDHLVGTVPLLVDVPPEEPKLSCFRKNPLVNAICEWRPSSTPSPTTKAVLFAKKINTTNGKSDFQVPCQYSQQLKSFSCQVEILEGDKVYHIVSLCVANSVGSKSSHNEAFHSLKMVQPDPPANLVVSAIPGRPRWLKVSWQHPETWDPSYYLLQFQLRYRPVWSKEFTVLLLPVAQYQCVIHDALRGVKHVVQVRGKEELDLGQWSEWSPEVTGTPWIAEPRTTPAGILWNPTQVSVEDSANHEDQYESSTEATSVLAPVQE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, Flow Cytometry 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 3570 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title