Learn More
IL-7R alpha/CD127 Antibody (22F3D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-22376
This item is not returnable.
View return policy
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Immunohistochemistry (Paraffin), CyTOF | |
| Amino acids 141-237 of human CD127 were used as the immunogen for the antibody. | |
| 0.1 mg | |
| IgG2b λ |
| Western Blot 2-5 ug/ml, Flow Cytometry 1:100. Use reported by customer review, CyTOF-ready | |
| Protein G purified | |
| Purified |