Learn More
IL-7R alpha/CD127 Antibody (73N8F2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-25264
 View more versions of this product
Product Code. 18145932
  • 453.00 € / 0.10mg
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-7R alpha/CD127
Monoclonal
1.0 mg/ml
Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
IL7R
Amino acids 145-235 of mouse IL-7R CD127 were used as the immunogen for the antibody.
0.1 mg
Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific
3575
Mouse
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
73N8F2
Unconjugated
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Rat
Protein G purified
RUO
Primary
This product reacts with Mouse.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a κ