missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
199.00 € - 440.00 €
Specifications
| Antigen | IL-9 |
|---|---|
| Clone | 795908 |
| Dilution | Neutralization 1-5 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774137
![]() |
R&D Systems
MAB2092-SP |
25 μg |
199.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18751317
![]() |
R&D Systems
MAB2092 |
100 μg |
440.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-9 Monoclonal specifically detects IL-9 in Human samples. It is validated for Neutralization.Specifications
| IL-9 | |
| Neutralization 1-5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine P40, HP40, IL9, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
| IL9 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 795908 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| P15248 | |
| 3578 | |
| Human embryonic kidney cell line HEK293-derived human IL-9 Met1-Ile144 Accession # P15248 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-9 in direct ELISA. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title