missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL12RB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524181
This item is not returnable.
View return policy
Description
IL12RB2 Polyclonal Antibody for Western Blot, IHC (P), Flow
The protein encoded by this gene is a type I transmembrane protein identified as a subunit of the interleukin 12 receptor complex. The coexpression of this and IL12RB1 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The expression of this gene is up-regulated by interferon gamma in Th1 cells, and plays a role in Th1 cell differentiation. The up-regulation of this gene is found to be associated with a number of infectious diseases, such as Crohn's disease and leprosy, which is thought to contribute to the inflammatory response and host defense.
Specifications
| IL12RB2 | |
| Polyclonal | |
| Unconjugated | |
| IL12RB2 | |
| A930027I18Rik; Ifnm; IL-12 receptor subunit beta-2; IL-12R subunit beta-2; Il12rb2; IL-12RB2; IL-12R-beta-2; interferon response to microorganisms; interleukin 12 receptor subunit beta 2; interleukin 12 receptor, beta 2; interleukin-12 receptor beta-2 chain; interleukin-12 receptor subunit beta-2; RP11-102M16.1 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 3595 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q99665 | |
| IL12RB2 | |
| KLH conjugated synthetic peptide between 756-783 amino acids from the C-terminal region of human IL12_2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction