missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL17D Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148582
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.
Specifications
| IL17D | |
| Polyclonal | |
| Unconjugated | |
| IL17D | |
| IL17D | |
| E. coli-derived recombinant mouse IL-17D, Ala25-Arg205. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| AI462269; FLJ30846; IL17D; IL-17D; IL-22; IL-27; Il27a; IL-3R; Interleukin; interleukin 17D; interleukin 27A; interleukin-17D; Interleukin-27; UNQ3096/PRO21175 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 239114 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction