missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL1RA Recombinant Rabbit Monoclonal Antibody (23GB6145), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
186.00 € - 446.00 €
Specifications
| Antigen | IL1RA |
|---|---|
| Clone | 23GB6145 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087963
![]() |
Invitrogen™
MA552990 |
100 μL |
446.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088916
|
Invitrogen™
MA552991 |
20 μL |
186.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL1RA Recombinant Monoclonal antibody specifically detects IL1RA in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| IL1RA | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P18510, P25085, P25086 | |
| 16181, 3557, 60582 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6145 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Anakinra; CD121A; cytokine; D2S1473; DIRA; F630041P17Rik; ICIL-1RA; IL 1 receptor antagonist; IL1 inhibitor; il1 receptor; il-1 receptor; IL1 receptor antagonist; IL1F3; IL1R; il-1r; IL1RA; IL-1RA; IL1-RA; IL-1ra3; IL-1R-alpha; IL1RN; IL-1RN; IL1RN (IL1F3); interleukin 1 receptor antagonist; interleukin-1 receptor; interleukin-1 receptor antagonist; interleukin-1 receptor antagonist protein; interleukin-1 receptor antagonist secretory form; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1ra); IRAP; IRAP1; MGC10430; MVCD4; P80; type II interleukin-1 receptor antagonist | |
| A synthesized peptide derived from human IL1RA (120-177AA). | |
| Primary | |
| -20°C | |
| IL1RN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title