missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL3RA / CD123 (Acute Myeloid Leukemia Marker) Mouse, Clone: IL3RA/2065, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | IL3RA / CD123 (Acute Myeloid Leukemia Marker) |
|---|---|
| Clone | IL3RA/2065 |
| Concentration | 200μg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15970268
|
EnquireBio
3563-MSM3-P0 |
20 μg | |||||||
|
15980268
|
EnquireBio
3563-MSM3-P1 |
100 μg |
576.45 € 478.00 € / 100µg Save 98.45 € 17% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| IL3RA / CD123 (Acute Myeloid Leukemia Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human IL3RA protein fragment (apprx. aa 26-171) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| ∽70kDa |
| IL3RA/2065 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| 3563 | |
| IgG2a κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cell Surface and Cytoplasmic | |
| THP-1, RPMI-8226 or HDLM-2 cells. Tonsil, Lymph Node or Stomach |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title