missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ILF2 Polyclonal Antibody, Biotin
GREENER_CHOICE

Product Code. 17856202
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17856202 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17856202 Supplier Invitrogen™ Supplier No. PA5142269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_004506.2; NP_001254738.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000. This antibody is tested in PAR-CLIP.

Nuclear factor of activated T-cells is a transcription factor required for T-cell expression of the interleukin 2 gene. NFAT binds to a sequence in the interleukin 2 gene enhancer known as the antigen receptor response element 2. In addition, NFAT can bind RNA and is an essential component for encapsidation and protein priming of hepatitis B viral polymerase. NFAT is a heterodimer of 45 kDa and 90 kDa proteins, the smaller of which is the product of this gene. The encoded protein binds strongly to the 90 kDa protein and stimulates its ability to enhance gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILF2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ILF2
Gene Accession No. Q12905
Gene Alias 6230405A16Rik; Ab1-143; ILF2; interleukin enhancer binding factor 2; interleukin enhancer binding factor 2, 45kD; interleukin enhancer binding factor 2, 45kDa; interleukin enhancer-binding factor 2; Interleukin enhancer-binding factor 2 homolog; liver regeneration-related protein LRRG031; MGC8391; Nf45; nuclear factor of activated T-cells 45 kDa; nuclear factor of activated T-cells, 45-kDa; PRO3063; TEG-267; testis expressed gene 267; Tex261; Tex267; Unknown (protein for MGC:134524); wu:fb34h01; zgc:73084; zgc:77276
Gene Symbols ILF2
Host Species Goat
Immunogen Peptide with sequence C-EKAYEKPPEKKEG (aa 355-367).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3608
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.