missing translation for 'onlineSavingsMsg'
Learn More

Iloprost, MedChemExpress

Product Code. 16490366
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490366 2 mg 2mg
16453776 25 mg 25mg
16438298 10 mg 10mg
16414187 50 mg 50mg
16410738 5 mg 5mg
16498026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16490366 Supplier Medchem Express Supplier No. HYA0096
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Iloprost (ZK 36374; Ciloprost) is a prostacyclin (PGI2) analogue, involves in embryo development and inflammation improvement, and inhibits tumor metastasis. Iloprost can be used for peripheral vascular research.

Iloprost (ZK 36374

Specifications

Chemical Name or Material Iloprost
Physical Form Oil
CAS 78919-13-8
Color Colorless
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C22H32O4
Synonym Ciloprost ZK 36374
Solubility Information DMSO : 100 mg/mL (277.40 mM; Need ultrasonic)
SMILES O=C(O)CCC/C=C1C[C@@]2([H])C[C@@H](O)[C@H](/C=C/[C@@H](O)C(C)CC#CC)[C@@]2([H])C\1
Molecular Weight (g/mol) 360.49
Formula Weight 360.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.