missing translation for 'onlineSavingsMsg'
Learn More

Imatinib, MedChemExpress

Product Code. 16481519
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
1 g
5 g
Unit Size:
1g
200mg
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481519 1 g 1g
16441776 500 mg 500mg
16431186 200 mg 200mg
16459127 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481519 Supplier Medchem Express Supplier No. HY15463
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Imatinib (STI571) is an orally bioavailable tyrosine kinases inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR and c-kit kinase activity. Imatinib (STI571) works by binding close to the ATP binding site, locking it in a closed or self-inhibited conformation, therefore inhibiting the enzyme activity of the protein semicompetitively. Imatinib also is an inhibitor of SARS-CoV and MERS-CoV.

Imatinib (STI571) is an orally bioavailable tyrosine kinases inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR and c-kit kinase activity. Imatinib (STI571) works by binding close to the ATP binding site, locking it in a closed or self-inhibited conformation, therefore inhibiting the enzyme activity of the protein semicompetitively. Imatinib also is an inhibitor of SARS-CoV and MERS-CoV.

Specifications

Chemical Name or Material Imatinib
Physical Form Solid
CAS 152459-95-5
Color Light Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C29H31N7O
For Use With (Application) Cancer-Kinase/protease
Synonym STI571 CGP-57148B
Solubility Information DMSO : 12.5 mg/mL (25.32 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CN(CC1)CCN1CC2=CC=C(C(NC3=CC=C(C)C(NC4=NC(C5=CC=CN=C5)=CC=N4)=C3)=O)C=C2
Molecular Weight (g/mol) 493.6
Formula Weight 493.6
Percent Purity 99.4%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.