missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imatinib mesylate, 98%

Product Code. 15658715 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15658715 5 g 5g
15648715 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15658715 Supplier Thermo Scientific Chemicals Supplier No. 461080050
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 220127-57-1 | C30H35N7O4S | 589.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220127-57-1
Molecular Formula C30H35N7O4S
Molecular Weight (g/mol) 589.72
MDL Number MFCD04307699
InChI Key YLMAHDNUQAMNNX-UHFFFAOYSA-N
Synonym imatinib mesylate, gleevec, glivec, imatinib mesilate, imatinib methanesulfonate, imatinib mesylate sti571, imatinib monomesylate, unii-8a1o1m485b, imatinib, methanesulfonate salt
PubChem CID 123596
ChEBI CHEBI:31690
IUPAC Name methanesulfonic acid;4-[(4-methylpiperazin-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
SMILES CS(O)(=O)=O.CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1

Specifications

Melting Point 209.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 589.71
Percent Purity 98%
Chemical Name or Material Imatinib mesylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.