accessibility menu, dialog, popup

UserName

CH3NNOHNH3CHNNNNCH3SOHOO

CAS RN 220127-57-1

IUPAC Name: N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)-4-[(4-methylpiperazin-1-yl)methyl]benzamide; methanesulfonic acid
Synonyms: gleevec imatinib methanesulfonate glivec imatinib mesilate
Molecular Weight (g/mol): 589.72
Molecular Formula: C30H35N7O4S
InChi Key: YLMAHDNUQAMNNX-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (8)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (4)